Skip to Content
Merck
All Photos(2)

Key Documents

459739

Sigma-Aldrich

1-(3-Bromopropyl)-2,2,5,5-tetramethyl-1-aza-2,5-disilacyclopentane

97%

Synonym(s):

1-(3-Bromopropyl)-2,2,5,5-tetramethyl-1-aza-2,5-disilacyclopentane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€272.00

€272.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€272.00

About This Item

Empirical Formula (Hill Notation):
C9H22BrNSi2
CAS Number:
Molecular Weight:
280.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€272.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.484 (lit.)

bp

80 °C/0.5 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

C[Si]1(C)CC[Si](C)(C)N1CCCBr

InChI

1S/C9H22BrNSi2/c1-12(2)8-9-13(3,4)11(12)7-5-6-10/h5-9H2,1-4H3

InChI key

BKBUBAVZGMRPAK-UHFFFAOYSA-N

Application

1-(3-Bromopropyl)-2,2,5,5-tetramethyl-1-aza-2,5-disilacyclopentane may be used for the synthesis of PFS-b-PZLys [PFS = Polyferrocenylsilane, PZLys = Poly(epsilon-benzyloxycarbonyl-L-lysine)] block copolymers.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yishan Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(28), 8624-8631 (2008-08-01)
A new type of metallopolymer-polypeptide block copolymer poly(ferrocenyldimethylsilane)-b-poly (epsilon-benzyloxycarbonyl-L-lysine) was synthesized by ring-opening polymerization of epsilon-benzyloxycarbonyl-L-lysine N-carboxyanhydride initiated by using a primary amino-terminated poly(ferrocenyldimethylsilane). Studies on the self-organization behavior of this polypeptide block copolymer in both the bulk state and
NANOSTRUCTURES FROM OLIGOPEPTIDE-POLYMER CONJUGATES.
Frauenrath H and Tian L.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 50(2), 533-533 (2009)
Synthesis and Characterization of Poly (L-Aspartic Acid)-Based Chimeras using High Vacuum Techniques.
Messman JM, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 47(2), 172-172 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service