Skip to Content
Merck
All Photos(1)

Key Documents

455547

Sigma-Aldrich

Ferroceneboronic acid

Synonym(s):

(Boronocyclopentadienyl)cyclopentadienyl iron, NSC 119337

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BFeO2
CAS Number:
Molecular Weight:
229.85
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

reaction suitability

reagent type: catalyst
core: iron

mp

145-150 °C (dec.) (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.OB(O)[C]2[CH][CH][CH][CH]2

InChI

1S/C5H6BO2.C5H5.Fe/c7-6(8)5-3-1-2-4-5;1-2-4-5-3-1;/h1-4,7-8H;1-5H;

InChI key

HHGAJIKAUQWFKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Redox-acitce electrochemical probe to monitor interactions involving sugars

Reactant involved in:
  • Synthesis of ferrocene-appended subporphyrins for electrochemical property studies
  • Synthesis of catalysts for polymerization of olefins
  • Suzuki-coupling reaction with organic triflates
  • Catalytic ortho-lithiation

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service