Skip to Content
Merck
All Photos(3)

Key Documents

454311

Sigma-Aldrich

N-Z-Ethylenediamine hydrochloride

95%

Synonym(s):

N-Z-1,2-Diaminoethane hydrochloride, Benzyl N-(2-aminoethyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONHCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
230.69
Beilstein:
4724532
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

169-172 °C (lit.)

functional group

Cbz
amine

SMILES string

[H]Cl.NCCNC(OCC1=CC=CC=C1)=O

InChI

1S/C10H14N2O2.ClH/c11-6-7-12-10(13)14-8-9-4-2-1-3-5-9;/h1-5H,6-8,11H2,(H,12,13);1H

InChI key

QMLKQXIAPAAIEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service