Skip to Content
Merck
All Photos(1)

Key Documents

44979

Sigma-Aldrich

1-Methyl-3-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)imidazolium hexafluorophosphate

≥97.0% (NMR)

Synonym(s):

1-Methyl-3-(1H,1H,2H,2H-perfluorooctyl)imidazolium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F19N2P
CAS Number:
Molecular Weight:
574.16
Beilstein:
8749297
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0% (NMR)

mp

78-81 °C

SMILES string

F[P-](F)(F)(F)(F)F.Cn1cc[n+](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c1

InChI

1S/C12H10F13N2.F6P/c1-26-4-5-27(6-26)3-2-7(13,14)8(15,16)9(17,18)10(19,20)11(21,22)12(23,24)25;1-7(2,3,4,5)6/h4-6H,2-3H2,1H3;/q+1;-1

InChI key

ZBUXZWKNRXKPNV-UHFFFAOYSA-N

Physical form

ionic liquid

Other Notes

Fluoroalkyl imidazolium ionic liquid and surfactant allowing the use of perfluorinated compounds in ionic liquids[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.L. Merrigan et al.
Chemical Communications (Cambridge, England), 2051-2051 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service