Skip to Content
Merck
All Photos(1)

Key Documents

446270

Sigma-Aldrich

5-Ethyl-2,8-diisopropyl-1-aza-3,7-dioxabicyclo[3.3.0]octane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H25NO2
CAS Number:
Molecular Weight:
227.34
EC Number:
MDL number:
UNSPSC Code:
12162002

refractive index

n20/D 1.455 (lit.)

viscosity

30 cP(lit.)

bp

127 °C/15 mmHg (lit.)

solubility

polyols and organic solvents: soluble

density

0.955 g/mL at 25 °C (lit.)

SMILES string

CCC12COC(C(C)C)N1C(OC2)C(C)C

Application

Moisture-scavenger.
Reactive diluent for urethane prepolymers. Replaces polyol component.

Features and Benefits

Hydrolyzed by water to yield a ketone and amino alcohol, which reacts with isocyanate groups.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service