Skip to Content
Merck
All Photos(1)

Key Documents

443751

Sigma-Aldrich

2,2,3,3,4,4,4-Heptafluorobutyl acrylate

97%

Synonym(s):

HFBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CF2CF2CF3
CAS Number:
Molecular Weight:
254.10
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.331 (lit.)

bp

120-122 °C/743 mmHg (lit.)

density

1.418 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)COC(=O)C=C

InChI

1S/C7H5F7O2/c1-2-4(15)16-3-5(8,9)6(10,11)7(12,13)14/h2H,1,3H2

InChI key

PLXOUIVCSUBZIX-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Letícia Rocha Guidi et al.
Talanta, 163, 85-93 (2016-11-26)
The objective of this study was to develop and validate a fast, sensitive and simple liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) method for the screening of six classes of antibiotics (aminoglycosides, beta-lactams, macrolides, quinolones, sulfonamides and tetracyclines) in fish. Samples
David Palmer et al.
Drug testing and analysis, 9(10), 1499-1505 (2017-01-15)
Gonadotropin-releasing hormone (GnRH) is a 10-residue peptide hormone that induces secretion of luteinizing hormone (LH) and follicle-stimulating hormone into the blood from the pituitary gland. In males, LH acts on the testes to produce testosterone. The performance-enhancing potential of testosterone
Michael Hellwig et al.
Journal of agricultural and food chemistry, 65(17), 3532-3540 (2017-04-19)
As a unique feature, honey from the New Zealand manuka tree (Leptospermum scoparium) contains substantial amounts of dihydroxyacetone (DHA) and methylglyoxal (MGO). Although MGO is a reactive intermediate in the Maillard reaction, very little is known about reactions of MGO
Erika M Palmieri et al.
Antioxidants & redox signaling, 26(8), 351-363 (2016-11-03)
Microglial cells are brain-resident macrophages engaged in surveillance and maintained in a constant state of relative inactivity. However, their involvement in autoimmune diseases indicates that in pathological conditions microglia gain an inflammatory phenotype. The mechanisms underlying this change in the
Jiyoun Kim et al.
Journal of agricultural and food chemistry, 64(48), 9203-9213 (2016-12-10)
Epigallocatechin gallate (EGCG) is the most abundant catechin found in the leaves of green tea, Camellia sinensis. In this study, novel epigallocatechin gallate-glucocides (EGCG-Gs) were synthesized by using dextransucrase from Leuconostoc mesenteroides B-1299CB4. Response surface methodology was adopted to optimize

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service