Skip to Content
Merck
All Photos(1)

Key Documents

443743

Sigma-Aldrich

4-Bromo-1,1,2-trifluoro-1-butene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€60.90

€60.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€60.90

About This Item

Linear Formula:
BrCH2CH2CF=CF2
CAS Number:
Molecular Weight:
188.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€60.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.401 (lit.)

bp

95-98 °C (lit.)

density

1.639 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

F\C(F)=C(/F)CCBr

InChI

1S/C4H4BrF3/c5-2-1-3(6)4(7)8/h1-2H2

InChI key

GQCQMFYIFUDARF-UHFFFAOYSA-N

General description

4-Bromo-1,1,2-trifluoro-1-butene is a fluorinated building block. It reacts with potassium hydroxide in the presence of tetrabutylammonium bromide (a phase transfer catalyst) to afford 1,1,2-trifluoro-1,3-butadiene.[1]

Application

4-Bromo-1,1,2-trifluoro-1-butene may be used in the preparation of 1,1,2-trifluoro-1-butene, via reaction with KOH in the presence of phase-transfer catalyst, tetrabutylammonium bromide.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 1, 2-Trifluoro-1, 3-butadiene: a convenient C4 intermediate for functionalized monofluoroolefins.
Matsuo N and Kende AS.
The Journal of Organic Chemistry, 53(10), 2304-2308 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service