Skip to Content
Merck
All Photos(1)

Documents

442941

Sigma-Aldrich

4-(4-Methylphenyl)-4-oxobutyric acid

98%

Synonym(s):

3-p-Toluoylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH2CH2CO2H
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

127-130 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)CCC(O)=O

InChI

1S/C11H12O3/c1-8-2-4-9(5-3-8)10(12)6-7-11(13)14/h2-5H,6-7H2,1H3,(H,13,14)

InChI key

OEEUWZITKKSXAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(4-Methylphenyl)-4-oxobutyric acid is an organic building block. It is a commercial corrosion inhibitor.

Application

4-(4-Methylphenyl)-4-oxobutyric acid (3-(4-Methylbenzoyl)propionic acid) may be used for the synthesis of polymetallic iron(III) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frey et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(8), 1407-1415 (2000-06-07)
Two polymetallic iron(III) complexes 1 and 2 have been synthesised from the known corrosion inhibitor 3-(4-methylbenzoyl)-propionic acid HL1 and their crystal structures determined. Coordination geometries extracted from these structures have been used as the basis for molecular modelling onto idealised
Glen B Deacon et al.
Chemical communications (Cambridge, England), (23)(23), 2820-2821 (2002-12-14)
The syntheses and structures of the novel Ce-Fe bimetallic complexes [[Fe(sal)2(bpy)]2Ce(NO3)(H2O)3].EtOH and [[Fe(sal)2(bpy)]4Ce2(H2O)11][salH]2.EtOH.3H2O (salH2 = salicylic acid) suggest Fe(3+)-sal2- units and Ce-OC(R)O-Fe bridging contribute to the formation of corrosion inhibitive layers on steel surfaces exposed to [Ce(salH)3(H2O)].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service