442585
Pentafluorophenylmagnesium bromide solution
0.5 M in diethyl ether
Synonym(s):
(Perfluorophenyl)magnesium bromide, 2,3,4,5,6-Pentafluorophenylmagnesium bromide, Bromo(pentafluorophenyl)magnesium
Select a Size
Select a Size
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
reaction type: Grignard Reaction
concentration
0.5 M in diethyl ether
bp
34.6 °C
density
0.861 g/mL at 25 °C
functional group
fluoro
SMILES string
Fc1c(F)c(F)c([Mg]Br)c(F)c1F
InChI
1S/C6F5.BrH.Mg/c7-2-1-3(8)5(10)6(11)4(2)9;;/h;1H;/q;;+1/p-1
InChI key
AXQVMJIDNDRSFM-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-40.0 °F - closed cup
Flash Point(C)
-40 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service