Skip to Content
Merck
All Photos(1)

Key Documents

44210

Sigma-Aldrich

tert-Dodecylmercaptan

technical, ≥97.0% (RT)

Synonym(s):

tert-Dodecanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H25SH
CAS Number:
Molecular Weight:
202.40
Beilstein:
1738382
EC Number:
UNSPSC Code:
12352100

grade

technical

Assay

≥97.0% (RT)

bp

227-248 °C (lit.)

density

0.86 g/mL at 20 °C (lit.)

InChI

1S/C12H26S/c1-9(2)10(3,4)11(5,6)12(7,8)13/h9,13H,1-8H3

InChI key

YAJYJWXEWKRTPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

mixture of isomers (mainly C12); prepared by reaction of ′tetrapropylene′ with H2S

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service