Skip to Content
Merck
All Photos(1)

Documents

440426

Sigma-Aldrich

1,4-Bis(dicyclohexylphosphino)butane

Synonym(s):

1,4-butane-diylbis(dicyclohexylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2P(CH2)4P(C6H11)2
CAS Number:
Molecular Weight:
450.66
Beilstein:
3368850
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

99-104 °C (lit.)

functional group

phosphine

SMILES string

C1CCC(CC1)P(CCCCP(C2CCCCC2)C3CCCCC3)C4CCCCC4

InChI

1S/C28H52P2/c1-5-15-25(16-6-1)29(26-17-7-2-8-18-26)23-13-14-24-30(27-19-9-3-10-20-27)28-21-11-4-12-22-28/h25-28H,1-24H2

InChI key

WNZGLXFLSFWPMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service