Skip to Content
Merck
All Photos(1)

Key Documents

440078

Sigma-Aldrich

Phenyl phenylacetate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2C6H5
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

96%

form

solid

bp

158 °C/7 mmHg (lit.)

mp

40-42 °C (lit.)

SMILES string

O=C(Cc1ccccc1)Oc2ccccc2

InChI

1S/C14H12O2/c15-14(11-12-7-3-1-4-8-12)16-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

USVNNHYNCCJCCP-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular rearrangements. Part XX. Thermolysis of carboxylic acid esters.
Aly MM, et al.
Canadian Journal of Chemistry, 61(7), 1532-1535 (1983)
Photochemistry of phenyl phenylacetates adsorbed on pentasil andfaujasite zeolites.
Tung C-H and Ying Y-M.
J. Chem. Soc. Perkin Trans. II, 7, 1319-1322 (1997)
C D Carrington et al.
The Biochemical journal, 236(2), 503-507 (1986-06-01)
For the purpose of assessing the neurotoxic potential of organophosphorus compounds, it has been determined that paraoxon-preinhibited hen brain has both neurotoxicant (mipafox)-sensitive (neurotoxic esterase; NTE) and -insensitive esterase components. Several experiments designed to investigate the kinetic parameters governing the
Selectivity in the photo-Fries reaction of phenyl phenylacetates included in a Nafion membrane.
Tung C-H and Xu X-H.
Tetrahedron Letters, 40(1), 127-130 (1999)
L Haagen et al.
European journal of clinical chemistry and clinical biochemistry : journal of the Forum of European Clinical Chemistry Societies, 30(7), 391-395 (1992-07-01)
A new method for phenotyping human serum arylesterase (EC 3.1.1.2) is described and evaluated. The aromatic esters, phenyl acetate and 4-nitrophenyl acetate, were compared as substrates for spectrophotometric measurement of arylesterase activity. A method for arylesterase phenotyping, based upon inhibition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service