Skip to Content
Merck
All Photos(1)

Key Documents

439711

Sigma-Aldrich

Neodymium(III) bromide

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Neodymium tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NdBr3
CAS Number:
Molecular Weight:
383.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

mp

684 °C (lit.)

density

5.3 g/mL at 25 °C (lit.)

SMILES string

Br[Nd](Br)Br

InChI

1S/3BrH.Nd/h3*1H;/q;;;+3/p-3

InChI key

LBWLQVSRPJHLEY-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service