Skip to Content
Merck
All Photos(1)

Key Documents

438839

Sigma-Aldrich

1-(Phenylsulfonyl)pyrrole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2S
CAS Number:
Molecular Weight:
207.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

88-91 °C (lit.)

SMILES string

O=S(=O)(c1ccccc1)n2cccc2

InChI

1S/C10H9NO2S/c12-14(13,11-8-4-5-9-11)10-6-2-1-3-7-10/h1-9H

InChI key

PPPXRIUHKCOOMU-UHFFFAOYSA-N

General description

1-(Phenylsulfonyl)pyrrole is a heterocyclic building block. 1-(Phenylsulfonyl) group serves as N-blocking and directing group in various organic syntheses.

Application

1-(Phenylsulfonyl)pyrrole (1-phenylsulfonyl-1H-pyrrole) may be used in the synthesis of 1-(phenylsulfonyl)pyrrole-2-boronic acid, via lithiation of 1-(phenylsulfonyl)-pyrrole. It may be used for the synthesis of 1-phenylsulfonyl-1H-pyrrole-3-sulfonyl chloride derivatives, which affords sulfonamide derivatives by reaction with nitrogen nucleophiles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrole chemistry. XXVIII. Substitution reactions of 1-(phenylsulfonyl) pyrrole and some derivatives.
Anderson HJ, et al.
Canadian Journal of Chemistry, 63(4), 896-902 (1985)
Efficient sulfonation of 1-phenylsulfonyl-1H-pyrroles and 1-phenylsulfonyl-1H-indoles using chlorosulfonic acid in acetonitrile.
Janosik T, et al.
Tetrahedron, 62(8), 1669-1707 (2006)
Synthesis of 2-Aryl-1-(phenylsulfonyl) pyrroles.
Grieb JG and Ketcha DM.
Synthetic Communications, 25(14), 2145-2153 (1995)
Papireddy Kancharla et al.
The Journal of organic chemistry, 79(23), 11674-11689 (2014-11-08)
Facile and highly efficient synthetic routes for the synthesis of (S)- and (R)-23-hydroxyundecylprodiginines ((23S)-2, and (23R)-2), 23-ketoundecylprodiginine (3), and deuterium-labeled 23-hydroxyundecylprodiginine ([23-d]-2) have been developed. We demonstrated a novel Rieske oxygenase MarG catalyzed stereoselective bicyclization of (23S)-2 to premarineosin A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service