Skip to Content
Merck
All Photos(1)

Key Documents

437875

Sigma-Aldrich

Cobalt(II) acetate tetrahydrate

99.999% trace metals basis

Synonym(s):

Cobaltous acetate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Co · 4H2O
CAS Number:
Molecular Weight:
249.08
Beilstein:
3731397
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

core: cobalt

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Co]OC(C)=O

InChI

1S/2C2H4O2.Co.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

ZBYYWKJVSFHYJL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) acetate tetrahydrate has an octahedral structure, the central cobalt orbit is coordinated by four water molecules and two acetate ligands. Cobalt(II) acetate tetrahydrate becomes anhydrous by 140oC.

Application

Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structures of nickel acetate, Ni(CH3COO)2.4H2O, and cobalt acetate, Co(CH3COO)2.4H2O
Van Niekerk JN and Schoening FRL
Acta Crystallographica, 6, 609-612 (1953)
Jaynes, B.S. et al.
Inorganic Chemistry, 34, 5735-5735 (1995)
Reagent Chemicals null
Vijayan Thamilarasan et al.
European journal of medicinal chemistry, 89, 266-278 (2014-12-03)
Metal complexes of the type Mn(bpy)2(N3)2 (1), Co(bpy)2(N3)2·3H2O (2) and Zn2(bpy)2(N3)4 (3) (Where bpy = 2,2-bipyridine) have been synthesized and characterized by elemental analysis and spectral (FT-IR, UV-vis) studies. The structure of complexes (1-3) have been determined by single crystal X-ray diffraction
Kyu-Nam Jung et al.
Scientific reports, 5, 7665-7665 (2015-01-08)
Rechargeable metal-air batteries are considered a promising energy storage solution owing to their high theoretical energy density. The major obstacles to realising this technology include the slow kinetics of oxygen reduction and evolution on the cathode (air electrode) upon battery

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service