Skip to Content
Merck
All Photos(1)

Key Documents

436925

Sigma-Aldrich

5-(4-Bromophenyl)furfural

97%

Synonym(s):

5-(4-Bromophenyl)-2-furancarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7BrO2
CAS Number:
Molecular Weight:
251.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

152-155 °C (lit.)

functional group

aldehyde
bromo

SMILES string

Brc1ccc(cc1)-c2ccc(C=O)o2

InChI

1S/C11H7BrO2/c12-9-3-1-8(2-4-9)11-6-5-10(7-13)14-11/h1-7H

InChI key

QRTAOOSYSVHQGT-UHFFFAOYSA-N

General description

5-(4-Bromophenyl)furfural is an aromatic aldehyde. Oxone oxidation of 5-(4-bromophenyl)furfural has been reported to afford γ-keto carboxylic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin R Travis et al.
Organic letters, 5(7), 1031-1034 (2003-03-28)
[reaction: see text] A highly efficient, mild, and simple protocol is presented for the oxidation of aldehydes to carboxylic acids utilizing Oxone as the sole oxidant. Direct conversion of aldehydes in alcoholic solvents to their corresponding ester products is also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service