Skip to Content
Merck
All Photos(1)

Key Documents

436089

Sigma-Aldrich

2-Fluoro-6-(trifluoromethyl)benzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)CHO
CAS Number:
Molecular Weight:
192.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

156 °C (lit.)

density

1.432 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1cccc(c1C=O)C(F)(F)F

InChI

1S/C8H4F4O/c9-7-3-1-2-6(5(7)4-13)8(10,11)12/h1-4H

InChI key

FAKUGVHRTLCKHB-UHFFFAOYSA-N

General description

2-Fluoro-6-(trifluoromethyl)benzaldehyde participates in the synthesis of 2-amino-5-trifluoromethylquinazoline.[1]

Application

2-Fluoro-6-(trifluoromethyl)benzaldehyde may be employed for the synthesis of diaryl ether, 2-(3,5-dimethoxyphenoxy)-6-(trifluoromethyl)- benzonitrile[2] and substituted benzo[b]thiophene-2-carboxylates.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-Aminoquinazolines from ortho-Fluorobenzaldehydes.
Hynes JB and Campbell JP.
Journal of Heterocyclic Chemistry, 34(2), 385-387 (1997)
Synthesis of Phloroglucinol Monoaryl Ethers.
Sherwood AM, et al.
Synthesis, 44, 1208-1212 (2012)
Tetrahedron Letters, 33, 7499-7499 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service