Skip to Content
Merck
All Photos(2)

Key Documents

435287

Sigma-Aldrich

Poly(styrene-co-maleic acid), partial isobutyl ester

average Mv ~65,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH(CO2R)CH(CO2R)]y, R = H or -CH2CH(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mv ~65,000

solubility

aqueous base, alcohols, ketones and ethyl acetate: soluble

SMILES string

OC(=O)\C=C\C(O)=O.C=Cc1ccccc1.CC(C)COC(=O)\C=C\C(=O)OCC(C)C

General description

1:<1 mole ratio of styrene:maleic anhydride

Application

Binder, water resistance additive and adhesive additive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik C Gunther et al.
Cell reports, 26(1), 145-158 (2019-01-04)
Cellular prion protein (PrPC) binds the scrapie conformation of PrP (PrPSc) and oligomeric β-amyloid peptide (Aβo) to mediate transmissible spongiform encephalopathy (TSE) and Alzheimer's disease (AD), respectively. We conducted cellular and biochemical screens for compounds blocking PrPC interaction with Aβo.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service