Skip to Content
Merck
All Photos(1)

Key Documents

434884

Sigma-Aldrich

Poly(dimethylsiloxane), distearate terminated

average Mn ~2,300

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

waxy solid (soft)

mol wt

average Mn ~2,300

solubility

H2O: insoluble

density

0.99 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(O)O.CCCCCCCCCCCCCCCCCC(O)=O

Application

Hydrophobic, non-greasy, lubricating films. Dispersant for pigments.

Physical properties

Liquifies under slight pressure, highly lubricious.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service