Skip to Content
Merck
All Photos(1)

Documents

434779

Sigma-Aldrich

Polybutadiene

average Mn 1,530-2,070 by VPO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH=CHCH2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

peroxide number <= 10 mval/kg

form

viscous liquid

mol wt

average Mn 1,530-2,070 by VPO

composition

cis-1,4, 72%
trans-1,4, 27%
vinyl, 1%

refractive index

n20/D 1.523

iodine value

425‑475 g/100g

transition temp

softening point −66-−54 °C

density

0.9 g/mL at 25 °C (lit.)

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+

InChI key

IAQRGUVFOMOMEM-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of water soluble binders and electrical encapsulants.

Features and Benefits

Contributes high corrosion resistance to coatings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Deniz Turgut et al.
PloS one, 5(12), e15551-e15551 (2010-12-24)
In recent years, there is aroused interest in expressing complex systems as networks of interacting nodes. Using descriptors from graph theory, it has been possible to classify many diverse systems derived from social and physical sciences alike. In particular, folded
Alexandra Sperschneider et al.
ACS nano, 4(10), 5609-5616 (2010-09-17)
We report on the quasi in situ scanning force microscopy nanotomography which proved to be a key method to effectively obtain a three-dimensional (3D) microdomain structure of a complex ABC triblock morphology. As an example, we studied polybutadiene-block-poly(2-vinyl pyridine)-block-poly(tert-butyl methacrylate)
Mechanochemistry: Demonstrated leverage.
Roman Boulatov
Nature chemistry, 5(2), 84-86 (2013-01-25)
Aleer M Yol et al.
Journal of the American Society for Mass Spectrometry, 24(1), 74-82 (2012-11-30)
[M + Ag](+) ions from cyclic and linear polystyrenes and polybutadienes, formed by matrix-assisted laser desorption ionization (MALDI), give rise to significantly different fragmentation patterns in tandem mass spectrometry (MS(2)) experiments. In both cases, fragmentation starts with homolytic cleavage at
Hiroshi Yabu
Chemical communications (Cambridge, England), 47(4), 1196-1197 (2010-11-27)
Blue light-emitting Au nanoclusters were synthesized in a simple one-pot process via reflux of Au ions with amino-terminated poly(1,2-butadiene) in toluene. DLS results and photoemission spectra showed that Au(8) nanoclusters were successfully formed. Moreover, photo-patterned films containing Au(8) clusters were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service