Skip to Content
Merck
All Photos(1)

Documents

434035

Sigma-Aldrich

N,S-Diacetyl-L-cysteine methyl ester

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COSCH2CH(NHCOCH3)CO2CH3
CAS Number:
Molecular Weight:
219.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −36.0°, c = 1 in methanol

mp

97-100 °C (lit.)

SMILES string

COC(=O)[C@H](CSC(C)=O)NC(C)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Karg et al.
Journal of dermatological science, 1(1), 39-45 (1990-01-01)
Thiols are of great importance for the regulation of many cellular functions including metabolism, transport and cell protection. In this study the usefulness of L-cysteine methyl and octyl esters, of N,S-diacetyl-L-cysteine methyl ester and glutathione isopropyl ester as cellular cysteine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service