Skip to Content
Merck
All Photos(1)

Key Documents

432873

Sigma-Aldrich

Pentaammine(trifluoromethanesulfonato)osmium(III) triflate

Synonym(s):

Pentaammine(trifluoromethanesulfonato)osmium(III) trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF3SO3Os(NH3)5](O3SCF3)2
CAS Number:
Molecular Weight:
722.59
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

reaction suitability

reagent type: catalyst
core: osmium

mp

~276 °C (lit.)

SMILES string

N.N.N.N.N.[Os+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.5H3N.Os/c3*2-1(3,4)8(5,6)7;;;;;;/h3*(H,5,6,7);5*1H3;/q;;;;;;;;+3/p-3

InChI key

HWGAHFXMBKZTPE-UHFFFAOYSA-K

Application

Useful for the dearomatization of anilines,[1] and the stereoselective synthesis of cyclohexanediones by osmium(II)-promoted Michael additions to phenols.[2] Precursor to Fischer carbyne complexes.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spera, M.L. et al.
Journal of the American Chemical Society, 119, 12772-12772 (1997)
Organometallics, 15, 245-245 (1996)
Journal of the American Chemical Society, 116, 6581-6581 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service