Skip to Content
Merck
All Photos(1)

Documents

432164

Sigma-Aldrich

Gadolinium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Gadolinium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdF3
CAS Number:
Molecular Weight:
214.25
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤150.0 ppm Trace Rare Earth Analysis

SMILES string

F[Gd](F)F

InChI

1S/3FH.Gd/h3*1H;/q;;;+3/p-3

InChI key

TYIZUJNEZNBXRS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the synthesis of non-oxide glasses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mater. Sci., 32, 741-741 (1996)
Journal of Non-Crystalline Solids, 203, 127-127 (1996)
Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service