Skip to Content
Merck
All Photos(2)

Key Documents

432067

Sigma-Aldrich

Terbium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Terbium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TbF3
CAS Number:
Molecular Weight:
215.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤150.0 ppm Trace Rare Earth Analysis

SMILES string

F[Tb](F)F

InChI

1S/3FH.Tb/h3*1H;/q;;;+3/p-3

InChI key

LKNRQYTYDPPUOX-UHFFFAOYSA-K

Application

For the preparation of electroluminescent thin films and luminescent zinc sulfide.[1][2][3][4] For the preparation of electroluminescent thin films and luminescent zinc sulfide.[1][2][3][4]
Useful for the preparation of fluoride glasses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adams, R.M. et al.
Materials Research Society Symposia Proceedings. Materials Research Society, 378-378 (1995)
Samokhvalov, M.K.
Zh. Prikl. Spektrosk., 62, 182-182 (1995)
Chem. Abstr., 124, 159253k-159253k (1996)
Chem. Abstr., 123, 353533w-353533w (1995)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service