Skip to Content
Merck
All Photos(1)

Key Documents

429856

Sigma-Aldrich

2-Amino-6-fluorobenzonitrile

99%

Synonym(s):

6-Fluoroanthranilonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(F)CN
CAS Number:
Molecular Weight:
136.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

125-128 °C (lit.)

functional group

fluoro
nitrile

storage temp.

2-8°C

SMILES string

Nc1cccc(F)c1C#N

InChI

1S/C7H5FN2/c8-6-2-1-3-7(10)5(6)4-9/h1-3H,10H2

InChI key

IQUNZGOZUJITBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-6-fluorobenzonitrile can be prepared by treatment of 2,6- difluorobenzonitrile with ammonia, which involves displacement of one of the activated fluorine atoms.[1]

Application

2-Amino-6-fluorobenzonitrile, a medicinal chemistry intermediate, has been employed for the preparation of tacrine-related compounds.[2] It may be used for the synthesis of antifolate and antibacterial quinazoline derivatives.[3]
It may be used for the synthesis bis(4-oxoquinazolin-2-yl)pyridine and fluoro-containing quinazolin-4(1H)-ones.[4] It may be employed for the synthesis of the following novel huprines[5]:
  • 12-amino-3-chloro-6,7,10,11-tetrahydro-9-isopropyl-7,11-methanocycloocta[b]quinoline hydrochloride
  • 9-allyl-12-amino-3-chloro-6,7,10,11-tetrahydro-7,11-methanocycloocta[b]quinoline hydrochloride
  • 12-amino-1-fluoro-6,7,10,11-tetrahydro-9-isopropyl-7,11-methanocycloocta[b]quinoline hydrochloride
  • 9-allyl-12-amino-1-fluoro-6,7,10,11-tetrahydro-7,11-methanocycloocta[b]quinoline hydrochloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Medicinal Chemistry, 29, 205-205 (1994)
N V Harris et al.
Journal of medicinal chemistry, 33(1), 434-444 (1990-01-01)
A series of 5-substituted 2,4-diaminoquinazolines (3) has been synthesized and evaluated as inhibitors of the enzyme dihydrofolate reductase (DHFR) from both bacterial and mammalian sources. The best compounds (e.g. 53) show good activity against Escherichia coli DHFR, but there is
Synthesis of 5-and 7-fluoroquinazolin-4 (1H)-ones.
Layeva AA, et al.
Russian Chemical Bulletin, 56(9), 1821-1827 (2007)
N-(aminophenyl)oxamic acids and esters as potent, orally active antiallergy agents.
D H Klaubert et al.
Journal of medicinal chemistry, 24(6), 742-748 (1981-06-01)
Julien Defaux et al.
Bioorganic & medicinal chemistry, 19(5), 1702-1707 (2011-02-15)
A series of 19 huprines has been evaluated for their activity against cultured bloodstream forms of Trypanosoma brucei and Plasmodium falciparum. Moreover, cytotoxicity against rat myoblast L6 cells was assessed for selected huprines. All the tested huprines are moderately potent

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service