Skip to Content
Merck
All Photos(1)

Key Documents

429082

Sigma-Aldrich

Hexamethylbenzene

purified by sublimation, ≥99%

Synonym(s):

1,2,3,4,5,6-Hexamethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6(CH3)6
CAS Number:
Molecular Weight:
162.27
Beilstein:
1905834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

purified by

sublimation

bp

264 °C (lit.)

mp

164-166 °C (lit.)

SMILES string

Cc1c(C)c(C)c(C)c(C)c1C

InChI

1S/C12H18/c1-7-8(2)10(4)12(6)11(5)9(7)3/h1-6H3

InChI key

YUWFEBAXEOLKSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethylbenzene (HMB) is a methyl benzene derivative. Its overtone spectra in carbon tetrachloride solution shows doublet structure due to two types of configurationally inequivalent hydrogens. The heat of solution of crystalline HMB has been obtained at 25°C from which the heat of fusion has been deduced by extrapolation process. The internal motion of the methyl groups has been explained by empirical force field calculations. Polarized Raman spectra of HMB at room temperature have been analyzed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

On the gearing of methyl groups in hexamethylbenzene.
Iroff LD.
Journal of Computational Chemistry, 1(1), 76-80 (1980)
The Raman spectrum of hexamethylbenzene single crystal.
Girlando A and Pecile C.
Chemical Physics Letters, 20(5), 467-470 (1973)
Detailed features in the local mode overtone bands of ethane, neopentane, tetramethylbutane, and hexamethylbenzene.
Henry BR and Greenlay WRA.
J. Chem. Phys. , 72(10), 5516-5524 (1980)
The Heats of Solution of Hexamethylbenzene, Cetyl Alcohol, and Dicetyl in Related Liquids; Heats of Fusion by an Extrapolation Process.
Parks GS and Rowe RD.
J. Chem. Phys. , 14(9), 507-510 (1946)
Binyu Lu et al.
Environmental science and pollution research international, 22(12), 9090-9099 (2015-04-16)
Contamination levels and spatial and temporal distributions of six typical synthetic musks (SMs) in water and sediment of the Songhua River in Northeastern China were investigated. Experimental data for 72 water and 52 sediment samples collected at 29 sampling sites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service