Skip to Content
Merck
All Photos(1)

Documents

428310

Sigma-Aldrich

Polyphenylsulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(-C6H4-4-SO2C6H4-4-OC6H4-4-C6H4-4-O-)n
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

beads

melt index

14-20 g/10 min (400°C/2.16kg)

refractive index

n20/D 1.672 (lit.)

transition temp

Tg 208 °C (DSC)

density

1.29 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(cc1)-c2ccc(O)cc2.Oc3ccc(cc3)S(=O)(=O)c4ccc(O)cc4

Application

Medical devices requiring sterilization and electrical connectors and switches.

Features and Benefits

Outstanding color, transparency and impact strength to 200°C. Best overall chemical resistance of the commercial polysulfones.

Physical form

Thermoplastic transparent engineering resin.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Watanabe et al.
Polymers, 12(1) (2020-01-16)
The purpose of this study was to evaluate the physical and chemical properties of engineering plastics processed using supercritical CO2. First, we prepared disk-shaped test pieces via a general molding process, which were plasticized using supercritical CO2 at temperatures lower

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service