Skip to Content
Merck
All Photos(1)

Key Documents

427659

Sigma-Aldrich

3-Chloro-2,4,5,6-tetrafluoropyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5ClF4N
CAS Number:
Molecular Weight:
185.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

118-121 °C (lit.)

density

1.609 g/mL at 25 °C (lit.)

SMILES string

Fc1nc(F)c(Cl)c(F)c1F

InChI

1S/C5ClF4N/c6-1-2(7)3(8)5(10)11-4(1)9

InChI key

UXUZMYHSICIOQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-2,4,5,6-tetrafluoropyridine (3-chlorotetrafluoropyridine) is a fluoropyridine derivative. Its quantum mechanical calculations of energies, geometries and vibrational wave numbers have been performed by DFT level of theory. The interpretation of its FT-IR and FT-Raman spectra have been reported. It forms corresponding organo-zinc compound by reacting with zinc. 3-Chlorotetrafluoropyridine reacts with tris(diethylamino)phosphine (P(NEt)3) in the presence of a proton donor to form product with fluorine replaced by hydrogen at position 4.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of polyfluoroarenes with zinc: hydrogenolysis and synthesis of polyfluoroaromatic organozinc compounds.
Krasnov VI and Platonov VE.
Journal of Fluorine Chemistry, 58(2), 247-247 (1992)
3-Fluoropyridyl nickel complexes as useful tools for the selective synthesis of new 2,4,5,6-tetrafluoropyridines: a route complementing the established methods to access fluorinated pyridines.
Sladek MI, et al.
New. J. Chem., 27(2), 313-318 (2003)
Heterocyclic polyfluoro-compounds. Part XII. Synthesis and some reactions of 2,3,5,6-tetrafluoro-4-iodopyridine.
Banks RE, et al.
J. Chem. Soc. Sect. C, 2091-2095 (1967)
Photochemical reaction of ethylene with 3-chlorotetrafluoropyridine: a novel insertion into a C-Cl bond.
Barlow MG, et al.
Journal of the Chemical Society. Chemical Communications, 14, 608-609 (1979)
Reactions of polyfluoroaromatic compounds with electrophilic agents in the presence of tris(dialkylamino)phosphines.
Bardin VV.
Russian Chemical Bulletin, 46(8), 1434-1436 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service