426962
Poly(4-ethylstyrene-co-divinylbenzene)
mean pore size 100 Å, 300-1200 μm particle size
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
beads
matrix
Poly(4-ethylstyrene-co-divinylbenzene)
particle size
300-1200 μm
pore size
100 Å mean pore size
55 % porosity
density
0.54 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
General description
Contains no functional groups. Polymeric adsorbent for removing soluble organics from polar solvents. Fast adsorption/desorption kinetics for large organic molecules. Resists degradation up to 150°C. Methanol is a good regenerant.
Application
Porous beads for adsorbing various soluble organics from polar solvents. Separates aromatic compounds from hydroxylic solvents such as aliphatic alcohols. Steroids and emulsifiers are also adsorbed.
Features and Benefits
Contains no functional groups. Resists degradation up to 150°C. Fast adsorption/desorption kinetics for large organic molecules. Methanol is a good regenerant.
Physical form
Hard porous polymeric adsorbent beads.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service