Skip to Content
Merck
All Photos(3)

Key Documents

425265

Sigma-Aldrich

6-tert-Butyl-2,3-naphthalenedicarbonitrile

94%

Synonym(s):

BRD9876

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC10H5(CN)2
CAS Number:
Molecular Weight:
234.30
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

94%

form

powder, crystals or chunks

impurities

<6% 4-tert-butyl-1,2-bis(dibromomethyl)benzene

mp

185-189 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)(C)c1ccc2cc(C#N)c(cc2c1)C#N

InChI

1S/C16H14N2/c1-16(2,3)15-5-4-11-6-13(9-17)14(10-18)7-12(11)8-15/h4-8H,1-3H3

InChI key

MKILROQBJOOZKC-UHFFFAOYSA-N

Application

6-tert-Butyl-2,3-naphthalenedicarbonitrile has been used for the preparation of tetra-t-butyl silicon naphthalocyanine bisoleate (SiNcBOA) suitable for LDL reconstitution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service