Skip to Content
Merck
All Photos(1)

Documents

424420

Sigma-Aldrich

Doyle dirhodium catalyst, Rh2(5S-MEPY)4

Synonym(s):

Dirhodium(II)tetrakis[methyl 2-pyrrolidone-5(S)-carboxylate], acetonitrile/2-propanol complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N4O12Rh2
CAS Number:
Molecular Weight:
774.34
UNSPSC Code:
12352103

Application

Chiral catalyst used for highly enantioselective intramolecular cyclopropanations, intermolecular addition reactions, and intramolecular carbon-hydrogen insertion reactions. Also employed in the synthesis of lactones and lactams in high diastereo-, regio-, and enantioselectivities. For a recent review, see Aldrichimica Acta

Other Notes

For research purposes only-Not for Use in Humans or in Human Clinical Trials.
Under Special Agreement with Regis Technologies, Inc.

Legal Information

Sold under license from Regis Technologies, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doyle, M.P.
Aldrichimica Acta, 29, 3-3 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service