421235
1-(2-Pyrimidyl)piperazine
98%
Synonym(s):
2-(1-Piperazinyl)pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H12N4
Molecular Weight:
164.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.587 (lit.)
bp
277 °C (lit.)
density
1.158 g/mL at 25 °C (lit.)
SMILES string
C1CN(CCN1)c2ncccn2
InChI
1S/C8H12N4/c1-2-10-8(11-3-1)12-6-4-9-5-7-12/h1-3,9H,4-7H2
InChI key
MRBFGEHILMYPTF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1-(2-Pyrimidyl)piperazine is a piperazine-based derivative. It is a metabolite of buspirone.
Application
1-(2-Pyrimidyl)piperazine may be used for the following studies:
- As derivatization reagent for the carboxyl groups on peptides.
- Carboxy group derivatization during the spectrophotometric analysis of phosphopeptides.
- Starting reagent for the synthesis of 3-{(4-(pyrimidin-2-yl)piperazin-1-yl)methyl}-1H-pyrrolo[2,3-b]pyridine.
- Synthesis of 3-phenyl-6-(4-(pyrimidin-2-yl)piperazin-1-yl)pyridazin-4-ol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Hascoët et al.
Pharmacology, biochemistry, and behavior, 67(1), 45-53 (2000-12-13)
Although numerous animal procedures have been employed in the study of antidepressants (ADs) in anxiety, the results following acute administration remain highly variable. The present study investigated the effect of the SSRI paroxetine (4, 8, and 16 mg/kg, IP) in
Xiaoqiang Qiao et al.
Rapid communications in mass spectrometry : RCM, 25(5), 639-646 (2011-02-04)
Piperazine-based derivatives, including 1-(2-pyridyl)piperazine (2-PP), 1-(2-pyrimidyl)piperazine (2-PMP), 1-(4-pyridyl)piperazine (4-PP), and 1-(1-methyl-4-piperidinyl)piperazine (M-PP), were used for the derivatization of carboxyl groups on peptides with 1-(3-dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride (EDC) and 1-hydroxy-7-azabenzotriazole (HOAt) as coupling reagents, and trifluoroacetic acid (TFA) as activator. Taking synthetic
A J Gower et al.
European journal of pharmacology, 155(1-2), 129-137 (1988-10-11)
The anxiolytic effects of buspirone, its metabolite, 1-(2-pyrimidyl)piperazine (1-PP) and several alpha 2-adrenoceptor antagonists have been compared in an anticonflict (shock-induced suppression of drinking) paradigm in rats. Idazoxan, WY 26392 and yohimbine had anticonflict effects comparable to those of buspirone
J Odontiadis et al.
Journal of pharmaceutical and biomedical analysis, 14(3), 347-351 (1996-01-01)
Buspirone is a member of the azapirone group of anxiolytic drugs and has one major metabolite, 1-(2-pyrimidinyl)piperazine (1-PP). The analyte, its metabolite and the internal standard were extracted from plasma utilizing solid-phase extraction columns. Chromatography was performed using isocratic reversed-phase
K T Kivistö et al.
Pharmacology & toxicology, 84(2), 94-97 (1999-03-06)
The effects of inhibition and induction of the metabolism of buspirone on the plasma concentrations of 1-(2-pyrimidinyl)-piperazine (a piperazine metabolite), the principal active metabolite of buspirone, were investigated. Two separate randomized, placebo-controlled cross-over studies with two phases were carried out
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service