All Photos(1)
About This Item
Linear Formula:
FC6H3(OH)CN
CAS Number:
Molecular Weight:
137.11
Beilstein:
4741066
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
EC Index Number:
422-810-7
Recommended Products
Assay
99%
mp
123-125 °C (lit.)
SMILES string
Oc1ccc(C#N)c(F)c1
InChI
1S/C7H4FNO/c8-7-3-6(10)2-1-5(7)4-9/h1-3,10H
InChI key
REIVHYDACHXPNH-UHFFFAOYSA-N
General description
2-Fluoro-4-hydroxybenzonitrile is a planar molecule. Its molecules are arranged in linear chains via weak intermolecular O-HN hydrogen bonds.
Application
2-Fluoro-4-hydroxybenzonitrile may be used in the synthesis of 4-cyano-3-fluorophenyl 4-(hexadecyloxy) benzoate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
2-Fluoro-4-hydroxybenzonitrile.
Song X-Y, et al.
Acta Crystallographica Section E, Structure Reports Online, 60(11), 1919-1920 (2004)
M K Usha et al.
Acta crystallographica. Section E, Structure reports online, 70(Pt 3), o244-o244 (2014-04-26)
In the title compound, C30H40FNO3, the dihedral angle between the benzene rings is 57.76 (7)°. The alkyl chain adopts an all-trans conformation. In the crystal, mol-ecules are linked by pairs of C-H⋯O hydrogen bonds, forming inversion dimers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service