Skip to Content
Merck
All Photos(1)

Key Documents

416886

Sigma-Aldrich

Acetic acid-d4

≥99.5 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Acetic-d3 acid-d, Tetradeuteroacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€320.00

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€320.00

About This Item

Linear Formula:
CD3CO2D
CAS Number:
Molecular Weight:
64.08
Beilstein:
1748971
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor density

2.07 (vs air)

vapor pressure

11.4 mmHg ( 20 °C)

isotopic purity

≥99.5 atom % D

Assay

≥99.00%

form

liquid

autoignition temp.

800 °F

contains

0.03 % (v/v) TMS

expl. lim.

16 %

technique(s)

NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.368 (lit.)

bp

115.5 °C (lit.)

mp

15-16 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]OC(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD

InChI key

QTBSBXVTEAMEQO-GUEYOVJQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid-d4 (deuterated acetic acid) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. Its dissociation constant in deuterium oxide has been obtained over a range of temperature by emf method.[1]

Application

Acetic acid-d4 may be used in the synthesis of deuterated (7E, 9Z) CLA (conjugated linoleic acid).[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dissociation of Acetic Acid-d4 in Deuterium Oxide from 5 to 50? and Related Isotope Effects.
Paabo M, et al.
The Journal of Physical Chemistry, 70(7), 2073-2077 (1966)
Stereocontrolled synthesis of (7E, 9Z)-[9, 10-2H]-conjugated linoleic acid.
Broustal G and Loreau O.
Journal of Labelled Compounds & Radiopharmaceuticals, 47(12), 875-880 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service