Skip to Content
Merck
All Photos(1)

Key Documents

415650

Sigma-Aldrich

3-(Dimethylamino)propyl acrylate

95%

Synonym(s):

3-(N ,N -Dimethylamino)propyl acrylate, N ,N -Dimethyl-3-aminopropyl acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€473.00

€473.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€473.00

About This Item

Linear Formula:
H2C=CHCO2(CH2)3N(CH3)2
CAS Number:
Molecular Weight:
157.21
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€473.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

167 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)CCCOC(=O)C=C

InChI

1S/C8H15NO2/c1-4-8(10)11-7-5-6-9(2)3/h4H,1,5-7H2,2-3H3

InChI key

UFQHFMGRRVQFNA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service