Skip to Content
Merck
All Photos(1)

Documents

415456

Sigma-Aldrich

1,2-Dichlorotetramethyldisilane

95%

Synonym(s):

1,1,2,2-Tetramethyl-1,2-dichlorodisilane, 1,2-Dichloro-1,1,2,2-tetramethyldisilane, Dichlorotetramethyldisilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[ClSi(CH3)2]2
CAS Number:
Molecular Weight:
187.22
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

148-149 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)[Si](C)(C)Cl

InChI

1S/C4H12Cl2Si2/c1-7(2,5)8(3,4)6/h1-4H3

InChI key

SFAZXBAPWCPIER-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service