Skip to Content
Merck
All Photos(3)

Key Documents

415340

Sigma-Aldrich

2-Amino-4′-bromoacetophenone hydrochloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCH2NH2·HCl
CAS Number:
Molecular Weight:
250.52
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

285 °C (dec.) (lit.)

functional group

amine
bromo
ketone

SMILES string

Cl.NCC(=O)c1ccc(Br)cc1

InChI

1S/C8H8BrNO.ClH/c9-7-3-1-6(2-4-7)8(11)5-10;/h1-4H,5,10H2;1H

InChI key

ROAVTVXTYFSQEA-UHFFFAOYSA-N

General description

2-Amino-4′-bromoacetophenone hydrochloride is an organic building block useful for chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service