Skip to Content
Merck
All Photos(1)

Key Documents

409553

Sigma-Aldrich

Calcium sulfide

−325 mesh, ≥99.9% trace metals basis

Synonym(s):

Calcium monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaS
CAS Number:
Molecular Weight:
72.14
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: calcium

particle size

−325 mesh

SMILES string

S=[Ca]

InChI

1S/Ca.S

InChI key

JGIATAMCQXIDNZ-UHFFFAOYSA-N

General description

Crystal structure of CaS is cubic.[1] Electrical conductivity of calcium sufide in the temperature range of 650oC-1000oC was studied in great detail. It was concluded that CaS could be used as a potential electrolyte in a sulfur concentration cell.2

Application

Calcium sulfide may be used to remove heavy metals from wastewater.[2]

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Chrysochoou et al.
Journal of hazardous materials, 179(1-3), 650-657 (2010-04-13)
Batch treatability studies for a Cr(VI)-contaminated glacial soil from a Cr plating facility were conducted using 1X and 2X the stoichiometric ratio of calcium polysulfide (CPS). The pH of the treated soil increased from 6 to 11 upon CPS addition
Masaaki Tabata et al.
Waste management (New York, N.Y.), 33(2), 356-362 (2012-12-04)
Dioxins like polychlorinated dibenzo-p-dioxins (PCSDDs), polychlorinated dibenzofurans (PCDFs) and polychlorinated biphenyls (PCBs) are mainly emitted from waste incinerators (WIs) and have become an international research focus because of its serious concerns over the adverse health effects. The detoxification of PCCDs/Fs
Kathryn L Bailey et al.
Ecotoxicology and environmental safety, 80, 195-202 (2012-03-27)
Foam delivery technology (FDT) uses surfactant based foam to immobilize subsurface contaminants in situ. Where traditional approaches are impractical, FDT has the potential to overcome many of the technical challenges facing the remediation of contaminated deep vadose zone environments. However
Masahiko Zuka et al.
Journal of forensic and legal medicine, 19(6), 358-362 (2012-08-01)
Lime sulfide poisoning by the oral route is rarely encountered in the practice of forensic science, whereas hydrogen sulfide poisoning is seen frequently. We report here two cases of fatal lime sulfide poisoning with several related cases and in addition
[Safety measure against improper use of lime sulfur].
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 23(2), 170-171 (2010-07-03)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service