Skip to Content
Merck
All Photos(1)

Key Documents

407690

Sigma-Aldrich

tert-Butyl N-(benzyloxy)carbamate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2ONHCO2C(CH3)3
CAS Number:
Molecular Weight:
223.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

99%

mp

45-47 °C (lit.)

functional group

amine
phenyl

SMILES string

CC(C)(C)OC(=O)NOCc1ccccc1

InChI

1S/C12H17NO3/c1-12(2,3)16-11(14)13-15-9-10-7-5-4-6-8-10/h4-8H,9H2,1-3H3,(H,13,14)

InChI key

MZNBNPWFHGWAGH-UHFFFAOYSA-N

General description

tert-Butyl N-(benzyloxy)carbamate (tert-butyl benzyloxycarbamate), a protected hydroxylamine,[1] is an N-alkyl-N-benzyloxy carbamate.[2] Its C-N cross coupling reaction with fluorescein ditriflate has been reported.[1] It participates in facile intramolecular cyclization with various carbon nucleophiles to afford functionalized 5- and 6-membered protected cyclic hydroxamic acids.[2]

Application

tert-Butyl N-(benzyloxy)carbamate was used in the preparation of seven-membered cyclic hydroxamic acids.[2] It may be used in the synthesis of 2-(N-formyl-N-hydroxyamino) ethylphosphonate (IPP).[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of rhodamines from fluoresceins using Pd-catalyzed C-N cross-coupling.
Grimm JB and Lavis LD.
Organic Letters, 13(24), 6354-6357 (2011)
Yuan Liu et al.
Tetrahedron, 67(12), 2206-2214 (2011-04-19)
N-Alkyl-N-benzyloxy carbamates, 2, undergo facile intramolecular cyclization with a variety of carbon nucleophiles to give functionalized 5- and 6-membered protected cyclic hydroxamic acids, 3, in good to excellent yields. This method can be extended to prepare seven-membered cyclic hydroxamic acids
I Kursula et al.
European journal of biochemistry, 268(19), 5189-5196 (2001-10-09)
The crystal structure of leishmania triosephosphate isomerase (TIM) complexed with 2-(N-formyl-N-hydroxy)-aminoethyl phosphonate (IPP) highlights the importance of Asn11 for binding and catalysis. IPP is an analogue of the substrate D-glyceraldehyde-3-phosphate, and it is observed to bind with its aldehyde oxygen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service