Skip to Content
Merck
All Photos(1)

Key Documents

40767

Sigma-Aldrich

(4S,5S)-(+)-2,2-Dimethyl-5-methylamino-4-phenyl-1,3-dioxane

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19NO2
CAS Number:
Molecular Weight:
221.30
Beilstein:
3591157
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D +43±2°, c = 1.5% in toluene

storage temp.

2-8°C

SMILES string

CN[C@H]1COC(C)(C)O[C@H]1c2ccccc2

InChI

1S/C13H19NO2/c1-13(2)15-9-11(14-3)12(16-13)10-7-5-4-6-8-10/h4-8,11-12,14H,9H2,1-3H3/t11-,12-/m0/s1

InChI key

OLDLOVICBLVDFQ-RYUDHWBXSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral auxiliary for the enantioselective synthesis of 4-keto esters

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Enders et al.
Synthesis, 659-659 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service