Skip to Content
Merck
All Photos(2)

Key Documents

405760

Sigma-Aldrich

Ethyl 2-oxo-1-cyclooctanecarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C8H13CO2C2H5
CAS Number:
Molecular Weight:
196.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.483 (lit.)

bp

85-87 °C/0.1 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CCCCCCC1=O

InChI

1S/C11H18O3/c1-2-14-11(13)9-7-5-3-4-6-8-10(9)12/h9H,2-8H2,1H3

InChI key

OJCDCHCBNASPBS-UHFFFAOYSA-N

General description

Ethyl 2-oxo-1-cyclooctanecarboxylate (2-carbethoxycyclooctanone) is a β-keto ester. It undergoes Michael addition with 3-buten-2-one (methyl vinyl ketone, MVK) in the presence of potassium bis(1,2-benzenediolato)phenylsilicate to afford the corresponding 1,4-adduct.[1] It can be synthesized from cycloheptanone.[2] Its boiling point, density and refractive index have been determined.[3]

Application

Ethyl 2-oxo-1-cyclooctanecarboxylate (2-carbethoxycyclooctanone) may be used in the synthesis of 2-carbethoxy-2-cyclooctenone.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated cyclooctenones are effective dienophiles.
Liu HJ, et al.
Canadian Journal of Chemistry, 75(6), 899-912 (1997)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 336-336 (2015)
Pentacoordinate Organosilicate-Catalyzed Michael Addition of ?-Keto Esters to 3-Buten-2-one.
Tateiwa J-U and Hosomi A.
European Journal of Organic Chemistry, 2001(8), 1445-1448 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service