All Photos(2)
About This Item
Linear Formula:
In(ClO4)3 · xH2O
CAS Number:
Molecular Weight:
413.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Recommended Products
grade
for analytical purposes
form
solid
composition
Degree of hydration, 8-10
reaction suitability
reagent type: catalyst
core: indium
reagent type: oxidant
concentration
≤27.2% In (EDTA titration)
SMILES string
O.[In+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
InChI
1S/3ClHO4.In.H2O/c3*2-1(3,4)5;;/h3*(H,2,3,4,5);;1H2/q;;;+3;/p-3
InChI key
BSUPSQJCOOBRFF-UHFFFAOYSA-K
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service