Skip to Content
Merck
All Photos(1)

Key Documents

401145

Sigma-Aldrich

Arsenic(III) iodide

Synonym(s):

Arsenic triiodide, Arsenous iodide, Triiodoarsine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AsI3
CAS Number:
Molecular Weight:
455.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 401145, please contact your local Merck office or dealer. Contact Technical Service

form

solid

reaction suitability

core: arsenic
reagent type: catalyst

density

4.69 g/mL at 25 °C (lit.)

SMILES string

I[As](I)I

InChI

1S/AsI3/c2-1(3)4

InChI key

IKIBSPLDJGAHPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service