Skip to Content
Merck
All Photos(1)

Documents

400971

Sigma-Aldrich

Calcium phosphide

Synonym(s):

Calcium Photophor, Tricalcium diphosphide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca3P2
CAS Number:
Molecular Weight:
182.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

chunks

particle size

1-7 mm

SMILES string

[Ca]=P[Ca]P=[Ca]

InChI

1S/3Ca.2P

InChI key

HMTIWQQJRUFNAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service