Skip to Content
Merck
All Photos(1)

Key Documents

400696

Sigma-Aldrich

Strontium iodide

AnhydroBeads, −10 mesh, ≥99.99% trace metals basis

Synonym(s):

Strontium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrI2
CAS Number:
Molecular Weight:
341.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

≥99.99% trace metals basis

form

beads

impurities

≤100.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

515 °C (lit.)

density

4.55 g/mL at 25 °C (lit.)

SMILES string

I[Sr]I

InChI

1S/2HI.Sr/h2*1H;/q;;+2/p-2

InChI key

KRIJWFBRWPCESA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service