Skip to Content
Merck
All Photos(1)

Key Documents

396729

Sigma-Aldrich

(2S,5S)-2,5-Hexanediol

99%, optical purity ee: 99% (GLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
118.17
Beilstein:
4652444
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +34.5°, c = 9 in chloroform

optical purity

ee: 99% (GLC)

bp

212-215 °C (lit.)

mp

50-53 °C (lit.)

SMILES string

C[C@H](O)CC[C@H](C)O

InChI

1S/C6H14O2/c1-5(7)3-4-6(2)8/h5-8H,3-4H2,1-2H3/t5-,6-/m0/s1

InChI key

OHMBHFSEKCCCBW-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of phospholanes. A building block in the synthesis of trans-2,5-dimethylpyrrolidine. C2 symmetric chiral diol with versatile applications as a chiral auxiliary, building block, and chiral ligand.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 2, 569-569 (1991)
The Journal of Organic Chemistry, 54, 1755-1755 (1989)
Stolle, A. et al.
Tetrahedron Letters, 35, 3521-3521 (1994)
Ho, O.C. et al.
Organometallics, 14, 2855-2855 (1995)
Marshall, J.A. Xie, S.
The Journal of Organic Chemistry, 60, 7230-7230 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service