Skip to Content
Merck
All Photos(1)

Key Documents

394998

Supelco

N,N-Dimethylformamide dipropyl acetal

for GC derivatization, LiChropur

Synonym(s):

1,1-Dipropoxy-N,N-dimethylmethylamine, 1,1-Dipropoxytrimethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

20 ML
€215.00

€215.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 ML
€215.00

About This Item

Linear Formula:
(CH3)2NCH(OCH2CH2CH3)2
CAS Number:
Molecular Weight:
175.27
Beilstein:
1098524
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

€215.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥97.5%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.409 (lit.)

bp

67-68 °C/14 mmHg (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

CCCOC(OCCC)N(C)C

InChI

1S/C9H21NO2/c1-5-7-11-9(10(3)4)12-8-6-2/h9H,5-8H2,1-4H3

InChI key

NSLGQFIDCADTAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide dipropyl acetal is an derivatizing reagent.[1]

Application

It was used as derivatizing agent in a study to determine cocaine and benzoyl ecgonine in urine using GC with on-column alkylation.[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N C Jain et al.
Journal of forensic sciences, 22(1), 7-16 (1977-01-01)
A gas chromatographic procedure has been developed for the simultaneous determination of cocaine and benzoyl ecgonine in urine specimens. The two drugs are extracted by isopropanol/chloroform from urine samples saturated with a bisalt buffer. The organic extract is evaporated to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service