Skip to Content
Merck
All Photos(1)

Key Documents

394629

Sigma-Aldrich

Reacta-Sil

Mixture of chlorotrimethylsilane, 1,1,1,3,3,3-hexamethyldisilazane, and pyridine (1:2:10)

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€226.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€226.00

About This Item

MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

density

0.938 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Cl.c1ccncc1.C[Si](C)(C)N[Si](C)(C)C

InChI

1S/C6H19NSi2.C5H5N.C3H9ClSi/c1-8(2,3)7-9(4,5)6;1-2-4-6-5-3-1;1-5(2,3)4/h7H,1-6H3;1-5H;1-3H3

InChI key

MNIBVWDUHGUQIQ-UHFFFAOYSA-N

Other Notes

Sediment is common and does not affect use

Legal Information

Reacta-Sil is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 제조번호: MKCF2195의 Quality release date: 2017-10-20 로 확인됩니다. 이번에 주문했을 때 'MKCF2195' 제품을 받았으며, 현재 유통되고 있는 Lot로 확인되는데 유효기한이 별도 명시가 되어 있지 않아 질문드립니다. 1. 유효기한은 별도 명시되어 있지 않은데 권장되는 유효기한이 있을까요 ? 2. 유효기한이 아예 없다면 근거 문서 또는 사유가 있을까요 ?

    1 answer
    1. This product does not contain an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service