Skip to Content
Merck
All Photos(1)

Key Documents

392588

Sigma-Aldrich

2,2,5,5-Tetramethyl-3-pyrrolidinecarboxamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O
CAS Number:
Molecular Weight:
170.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

128-131 °C (lit.)

SMILES string

CC1(C)CC(C(N)=O)C(C)(C)N1

InChI

1S/C9H18N2O/c1-8(2)5-6(7(10)12)9(3,4)11-8/h6,11H,5H2,1-4H3,(H2,10,12)

InChI key

POAGFQOGFRYOFM-UHFFFAOYSA-N

General description

2,2,5,5-Tetramethyl-3-pyrrolidinecarboxamide (2,2,5,5-Tetramethylpyrrolidine-3-carboxamide) is a carboxamide of hydrogenated pyrrole derivative. Its synthesis by the hydrogenation of 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide has been reported. The antiarrhythmic activity of some of the derivatives of 2,2,5,5-tetramethylpyrrolidine-3-carboxamide has been evaluated.

Application

2,2,5,5-Tetramethyl-3-pyrrolidinecarboxamide may be used in the synthesis of its nitroxide, 3-carbamoyl-2,2,5,5-tetramethyl-1-pyrrolidinyloxy free radical.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 260-260 (2015)
Nitroxide free radicals in the hydrogenated pyrrole series.
Rozantsev EG, et al.
Russian Chemical Bulletin, 15(4), 638-641 (1966)
O H Hankovszky et al.
Journal of medicinal chemistry, 29(7), 1138-1152 (1986-07-01)
N-(omega-Aminoalkyl)-2,2,5,5-tetramethyl-3-pyrroline- or -pyrrolidine-3-carboxamides were acylated on the primary amino group of the side chain by means of reactive acid derivatives (acid chlorides, activated esters, phthalic anhydrides, phthalimide, 2-alkyl-4H-3,1-benzoxazin-4-ones) or they were alkylated by forming the Schiff bases and subsequent sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service