Skip to Content
Merck
All Photos(1)

Key Documents

392200

Sigma-Aldrich

1,1′-Diethyl-4,4′-dicarbocyanine iodide

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27IN2
CAS Number:
Molecular Weight:
506.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 90%

mp

158 °C (dec.) (lit.)

λmax

814 nm

SMILES string

[I-].CCN1C=C\C(=C/C=C/C=C/c2cc[n+](CC)c3ccccc23)c4ccccc14

InChI

1S/C27H27N2.HI/c1-3-28-20-18-22(24-14-8-10-16-26(24)28)12-6-5-7-13-23-19-21-29(4-2)27-17-11-9-15-25(23)27;/h5-21H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

XDGZLJIBGBJNTI-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service